Description
- Speed: The luminescent format eliminates the need for time-consuming analyses such as HPLC and LC/MS
- Simplified Method: The simple 'add and read' protocol makes the assay amenable to higher throughput screening in multiwell plates
- Sensitive: Allows researchers to use less enzyme and scale down reaction volumes, which saves on reagent costs
Specifications
Specifications
| Content And Storage | Varies Among Components |
| Quantity | 1000 Assays |