Learn More
Promega UGT-Glo™ UGT2B7 Screening System
Catalog No. PRV2131 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRV2131 1000 Assays
1 options
Catalog No. PRV2131 Supplier Promega Supplier No. V2131
Provides a luminescent method for measuring UDP glucuronosyltransferase (UGT) activity. Signal generated is inversely correlated with UGT activity present in the sample.
  • Speed: The luminescent format eliminates the need for time-consuming analyses such as HPLC and LC/MS
  • Simplified Method: The simple 'add and read' protocol makes the assay amenable to higher throughput screening in multiwell plates
  • Sensitive: Allows researchers to use less enzyme and scale down reaction volumes, which saves on reagent costs
Specifications
Content And Storage Varies Among Components
Quantity 1000 Assays