Learn More
Invitrogen™ UGT1A1 Polyclonal Antibody

Description
Immunogen sequence: MAVESQGGRP LVLGLLLCVL GPVVSHAGKI LLIPVDGSHW LSMLGAIQQL QQRGHEIVVL APDASLYIRD GAFYTLKTYP VPFQREDVKE SFVSLGHNVF ENDSFLQRVI KTYKKIKKDS AMLLSGCSHL LHNKELMASL AESSFDVMLT DPFLPCSPIV AQYLSLPTVF FLHALPCSLE FEATQCPNPF SYVPRPLSSH; Positive Samples: HepG2, HL-60, SH-SY5Y, SW480, mouse liver, mouse kidney, mouse lung, rat liver; Cellular Location: Endoplasmic reticulum, Endoplasmic reticulum membrane, Microsome, Single-pass membrane protein.
Specifications
Specifications
| Antigen | UGT1A1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.82 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | UGT1A1 |
| Gene Accession No. | P22309, Q63886, Q64550 |
| Gene Alias | B1; BILIQTL1; bilirubin UDP-glucuronosyltranserase; bilirubin UDP-glucuronosyltransferase 1-1; bilirubin UDP-glucuronosyltransferase isozyme 1; bilirubin-specific UDPGT isozyme 1; EC 2.4.1.17; GNT1; HUG BR1; HUGBR1; hUG-BR1; UDP glucuronosyltransferase 1 family, polypeptide A1; UDP glucuronosyltransferase family 1 member A1; UDP glycosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1 family member 1; UDP-glucuronosyltransferase 1 family, member 1; UDP-glucuronosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1-A; UDP-glucuronosyltransferase 1A1; UDPGT; UDPGT 1-1; Udpgt-1a; UGT 1A; UGT1; UGT1 01; UGT1*1; UGT1.1; UGT1-01; UGT1A; UGT-1A; UGT1A01; UGT1A1; UGTBR1 |
| Gene Symbols | UGT1A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.