Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT1A1 Recombinant Rabbit Monoclonal Antibody (8U9P3)
GREENER_CHOICE

Catalog No. FEMA551266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551266 Supplier Invitrogen™ Supplier No. MA551266
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ENDSFLQRVI KTYKKIKKDS AMLLSGCSHL LHNKELMASL AESSFDVMLT DPFLPCSPIV AQYLSLPTVF FLHALPCSLE FEATQCPNPF SYVPRPLSSH.

This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. The preferred substrate of this enzyme is bilirubin, although it also has moderate activity with simple phenols, flavones, and C18 steroids. Mutations in this gene result in Crigler-Najjar syndromes types I and II and in Gilbert syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT1A1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8U9P3
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene UGT1A1
Gene Accession No. P22309, Q63886, Q64550
Gene Alias B1; BILIQTL1; bilirubin UDP-glucuronosyltranserase; bilirubin UDP-glucuronosyltransferase 1-1; bilirubin UDP-glucuronosyltransferase isozyme 1; bilirubin-specific UDPGT isozyme 1; EC 2.4.1.17; GNT1; HUG BR1; HUGBR1; hUG-BR1; UDP glucuronosyltransferase 1 family, polypeptide A1; UDP glucuronosyltransferase family 1 member A1; UDP glycosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1 family member 1; UDP-glucuronosyltransferase 1 family, member 1; UDP-glucuronosyltransferase 1 family, polypeptide A1; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1-A; UDP-glucuronosyltransferase 1A1; UDPGT; UDPGT 1-1; Udpgt-1a; UGT 1A; UGT1; UGT1 01; UGT1*1; UGT1.1; UGT1-01; UGT1A; UGT-1A; UGT1A01; UGT1A1; UGTBR1
Gene Symbols UGT1A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 101-200 of human UGT1A1 (NP_000454.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24861, 394436, 54658
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.