Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT1A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576752 Supplier Invitrogen™ Supplier No. PA576752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. This enzyme has some glucuronidase activity towards bilirubin, although is is more active on amines, steroids, and sapogenins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT1A4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene UGT1A4
Gene Accession No. P22310
Gene Alias bilirubin UDP-glucuronosyltransferase isozyme 2; bilirubin-specific UDPGT isozyme 2; GNT1; hUG-BR2; UDP glucuronosyltransferase 1 family, polypeptide A4; UDP glucuronosyltransferase family 1 member A4; UDP glycosyltransferase 1 family, polypeptide A4; UDP-glucuronosyltransferase 1 family polypeptide A4s; UDP-glucuronosyltransferase 1-4; UDP-glucuronosyltransferase 1A4; UDP-glucuronosyltransferase 1-D; UDPGT; UDPGT 1-4; UGT1; UGT1.4; UGT1-04; UGT1A4; UGT1A4S; UGT1D; UGT-1D
Gene Symbols UGT1A4
Host Species Rabbit
Immunogen Recombinant full length Human UGT1A4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54657
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.