Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT1A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543353 Supplier Invitrogen™ Supplier No. PA543353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EENFFTLTTY AISWTQDEFD RLLLGHTQSF FETEHLLMKF SRRMAIMNNM Sequence homology: Human: 100%; Rat: 83%.

This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT1A5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UGT1A5
Gene Accession No. P35504
Gene Alias B5; GNT1; UDP glucuronosyltransferase 1 family, polypeptide A5; UDP glucuronosyltransferase family 1 member A5; UDP glycosyltransferase 1 family, polypeptide A5; UDP-glucuronosyltransferase 1 family polypeptide A5s; UDP-glucuronosyltransferase 1-5; UDP-glucuronosyltransferase 1A5; UDP-glucuronosyltransferase 1-E; UDPGT; UDPGT 1-5; Ugt1; UGT1*5; UGT1.5; UGT1-05; UGT1A5; Ugt1a6; UGT1E; UGT-1E
Gene Symbols UGT1A5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human UGT1A5 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54579
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.