Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UGT1A5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2843220.1ML
Description
UGT1A5 Polyclonal specifically detects UGT1A5 in Human samples. It is validated for Western Blot.Specifications
UGT1A5 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54579 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
GNT1, polypeptide A5, UDP glucuronosyltransferase 1 family, polypeptide A5, UDP-glucuronosyltransferase 1-E, UDPGT 1-5, UGT1, UGT1*5, UGT1E, UGT-1E | |
The immunogen is a synthetic peptide directed towards the middle region of human UGT1A5 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction