Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT1A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116104 Supplier Thermo Scientific Supplier No. PA5116104
Add to Cart
Add to Cart

UGT1A6 Polyclonal Antibody for Western Blot, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT1A6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene UGT1A6
Gene Accession No. P08430, P19224, Q64435
Gene Alias A1; GNT1; HLUGP; HLUGP1; MGC29860; Phenol UDP-glucuronosyltransferase; phenol-metabolizing UDP-glucuronosyltransferase; P-nitrophenol specific; P-nitrophenol-specific UDPGT; UDP glucuronosyltransferase 1 family, polypeptide A6; UDP glucuronosyltransferase 1 family, polypeptide A6A; UDP glucuronosyltransferase 1A6; UDP glucuronosyltransferase family 1 member A6; UDP glycosyltransferase 1 family polypeptide A4; UDP glycosyltransferase 1 family, polypeptide A6; UDP-glucuronosyltransferase; UDP-glucuronosyltransferase 1 family polypeptide A6s; UDP-glucuronosyltransferase 1-6; UDP-glucuronosyltransferase 1A6; UDP-glucuronosyltransferase 1-F; Udpgt; UDPGT 1-6; UGP1A1; UGT1; UGT1*6; UGT1.6; UGT1-06; Ugt1a6; Ugt1a6a; UGT1A6S; UGT1A7; UGT1F; UGT-1F
Gene Symbols UGT1A6, Ugt1a6a
Host Species Rabbit
Immunogen A synthesized peptide derived from human UGT1A6(Accession P19224), corresponding to amino acid residues V60-T110.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113992, 54578, 94284
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.