Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT1A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543352 Supplier Invitrogen™ Supplier No. PA543352
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VKTYSTSYTL EDQDREFMVF ADARWTAPLR SAFSLLTSSS NGIFDLFFSN Sequence homology: Human: 100%.

UGT1A7 is an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. The enzyme encoded by this gene has moderate glucuronidase activity with phenols.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT1A7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UGT1A7
Gene Accession No. Q9HAW7
Gene Alias GNT1; UDP glucuronosyltransferase 1 family, polypeptide A7; UDP glucuronosyltransferase family 1 member A7; UDP glycosyltransferase 1 family, polypeptide A7; UDP-glucuronosyltransferase 1 family polypeptide A7s; UDP-glucuronosyltransferase 1-7; UDP-glucuronosyltransferase 1A7; UDP-glucuronosyltransferase 1-G; UDPGT; UDPGT 1-7; UGT1; UGT1.7; UGT1-07; UGT1A7; UGT1G; UGT-1G
Gene Symbols UGT1A7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human UGT1A7 (aa 74-123).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54577
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.