Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UGT1A9 Mouse anti-Human, Clone: 14G11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS222925UG
Description
UGT1A9 Monoclonal antibody specifically detects UGT1A9 in Human samples. It is validated for Immunocytochemistry.
Specifications
| UGT1A9 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| KLH-conjugated linear peptide corresponding to 32 amino acids from the N-terminal half of human UDP-glucuronosyltransferase 1-9 (UDT1A9). | |
| 25 μg | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. |
| Immunocytochemistry | |
| 14G11 | |
| Unconjugated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction