Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT1A9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985991
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5991 Supplier Bioss Supplier No. BS4224R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UGT1A9 is a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. UGT1A9 is active on phenols.This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5′ exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. The enzyme encoded by this gene is active on phenols. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Specifications

Antigen UGT1A9
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UGT1A9
Gene Accession No. O60656
Gene Alias GNT1; HLUGP4; lugP4; mUGTBr/P; SHEUGT1A07; UDP glucuronosyltransferase 1 family, polypeptide A9; UDP glucuronosyltransferase family 1 member A9; UDP glycosyltransferase 1 family polypeptide A10; UDP glycosyltransferase 1 family polypeptide A11; UDP glycosyltransferase 1 family polypeptide A12; UDP glycosyltransferase 1 family, polypeptide A10; UDP glycosyltransferase 1 family, polypeptide A9; UDP-glucuronosyltransferase; UDP-glucuronosyltransferase 1 family polypeptide A9s; UDP-glucuronosyltransferase 1-7; UDP-glucuronosyltransferase 1-9; UDP-glucuronosyltransferase 1A9; UDP-glucuronosyltransferase 1-I; UDPGT; UDPGT 1-9; UGT1; UGT1.9; UGT1-09; UGT1-9; Ugt1a10; Ugt1a11; Ugt1a12; Ugt1a9; UGT1A9S; UGT1AI; UGT1I; UGT-1I; UGTP4
Gene Symbols UGT1A9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human UGT1A9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54600
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.