Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UGT2A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169358
Description
UGT2A3 Polyclonal antibody specifically detects UGT2A3 in Human samples. It is validated for Western Blot.Specifications
UGT2A3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 2.4.1.17, FLJ21934, UDP glucuronosyltransferase 2 family, polypeptide A3, UDP-glucuronosyltransferase 2A3, UDPGT 2A3 | |
Rabbit | |
60 kDa | |
100 μL | |
Lipid and Metabolism | |
79799 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q6UWM9 | |
UGT2A3 | |
Synthetic peptides corresponding to UGT2A3(UDP glucuronosyltransferase 2 family, polypeptide A3) The peptide sequence was selected from the middle region of UGT2A3. Peptide sequence GIVVFSLGSLFQNVTEEKANIIASALAQIPQKVLWRYKGKKPSTLGANTR. | |
Affinity purified | |
RUO | |
Primary | |
Canine: 79%. | |
Human, Mouse, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction