Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT2A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548900
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548900 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548900 Supplier Invitrogen™ Supplier No. PA548900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UGT2A3 Polyclonal Antibody for Western Blot

UDP-glucuronosyltransferases catalyze phase II biotransformation reactions in which lipophilic substrates are conjugated with glucuronic acid to increase water solubility and enhance excretion. They are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT2A3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene UGT2A3
Gene Accession No. Q6UWM9
Gene Alias UDP glucuronosyltransferase 2 family, polypeptide A3; UDP glucuronosyltransferase family 2 member A3; UDP-glucuronosyltransferase 2A3; UDPGT 2A3; UGT2A3; UNQ2559/PRO6239
Gene Symbols UGT2A3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 324-353 amino acids from the Central region of human UGT2A3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.