Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UGT2B Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP172063
Description
UGT2B Polyclonal specifically detects UGT2B in Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
UGT2B | |
Polyclonal | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_113721 | |
UGT2B | |
Peptide with sequence C-KLDFLSALEEVIDN corresponding to internal region according to NP_113721.3. | |
0.1 mg | |
Primary | |
This antibody is NOT expected to cross-react with other Ugt2 proteins | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Ugt2b UDP glycosyltransferase 2 family, polypeptide B, Ugt2b2 | |
Goat | |
Affinity Purified | |
RUO | |
24862 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction