Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UGT2B15 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548363
Description
The UGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B8 demonstrates reactivity with estriol. The UGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B8 demonstrates reactivity with estriol. See UGT2B4 (MIM 600067).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Specifications
UGT2B15 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P54855 | |
UGT2B15 | |
KLH conjugated synthetic peptide between 156-185 amino acids from the Central region of human UGT2B15. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
UGT2B15 | |
UDP-glucuronosyltransferase 2B15, UDPGT 2B15, HLUG4, UDP-glucuronosyltransferase 2B8, UDPGT 2B8, UDPGTh-3, UGT2B15, UGT2B8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7366 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction