Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT2B4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560926 Supplier Invitrogen™ Supplier No. PA560926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVGGLHCKPA KPLPKEMEEF VQSSG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 96%.

UGT2B4 belongs to the UDP-glycosyltransferase family. UDPGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. This isozyme is active on polyhydroxylated estrogens (such as estriol, 4-hydroxyestrone and 2-hydroxyestriol) and xenobiotics (such as 4-methylumbelliferone, 1-naphthol, 4-nitrophenol, 2-aminophenol, 4-hydroxybiphenyl and menthol). It is capable of 6 alpha-hydroxyglucuronidation of hyodeoxycholic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT2B4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UGT2B4
Gene Accession No. P06133
Gene Alias HLUG25; hyodeoxycholic acid; hyodeoxycholic acid-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B15; UDP glucuronosyltransferase 2 family, polypeptide B36; UDP glucuronosyltransferase 2 family, polypeptide B4; UDP glucuronosyltransferase family 2 member B4; UDP glycosyltransferase 2 family, polypeptide B4; UDP-glucuronosyltransferase 2B12; UDP-glucuronosyltransferase 2B15; UDP-glucuronosyltransferase 2B36; UDP-glucuronosyltransferase 2B4; UDP-glucuronyltransferase, family 2, beta-4; UDPGT 2B12; UDPGT 2B15; UDPGT 2B36; UDPGT 2B4; UDPGT2B4; UDPGTH1; UDPGTh-1; UGT2B11; Ugt2b12; Ugt2b15; Ugt2b36; UGT2B4
Gene Symbols UGT2B4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UGT2B28. Recombinant protein control fragment (Product #RP-104381).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7363
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.