Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UGT2B7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529947
Description
Recommended positive controls: HepG2. Predicted reactivity: Sheep (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The UGTs serve a major role in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B7 has unique specificity for 3,4-catechol estrogens and estriol, suggesting that it may play an important role in regulating the level and activity of these potent estrogen metabolites. Its subcellular location is the microsome.
Specifications
UGT2B7 | |
Polyclonal | |
Unconjugated | |
UGT2B7 | |
3,4-catechol estrogen specific; 3,4-catechol estrogen-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B7; UDP glucuronosyltransferase 2B7; UDP glucuronosyltransferase family 2 member B7; UDP-glucuronosyltransferase 2B7; UDP-glucuronosyltransferase 2B8; UDP-glucuronosyltransferase 2B9; UDP-glucuronyltransferase, family 2, beta-7; UDPGT; UDPGT 2B7; UDPGT 2B8; UDPGT 2B9; UDPGT2B7; UDPGTh 2; UDPGTH2; UDPGTh-2; Ugt2b7; Ugt2b8; UGT2B9; UGT2B-RH4; UGTB2B9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7364 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P16662 | |
UGT2B7 | |
Recombinant protein fragment corresponding to a region within amino acids 178 and 466 of Human UGT2B7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction