Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT3A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543280 Supplier Invitrogen™ Supplier No. PA543280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WFSPEDHQKR IKKHFDSYIE TALDGRKESE ALVKLMEIFG TQCSYLLSRK Sequence homology: Human: 100%.

UDP-glucuronosyltransferases catalyze phase II biotransformation reactions in which lipophilic substrates are conjugated with glucuronic acid to increase water solubility and enhance excretion. They are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT3A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UGT3A1
Gene Accession No. Q6NUS8
Gene Alias FLJ26528; FLJ34658; UD3A1; UDP glycosyltransferase 3 family, polypeptide A1; UDP glycosyltransferase family 3 member A1; UDP-glucuronosyltransferase 3A1; UDPGT 3A1; UGT3A1
Gene Symbols UGT3A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human UGT3A1 (aa 79-128).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133688
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.