Learn More
Invitrogen™ UGT3A1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total UGT3A1 protein.
Specifications
Specifications
| Antigen | UGT3A1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | UGT3A1 |
| Gene Accession No. | Q6NUS8 |
| Gene Alias | FLJ26528; FLJ34658; UD3A1; UDP glycosyltransferase 3 family, polypeptide A1; UDP glycosyltransferase family 3 member A1; UDP-glucuronosyltransferase 3A1; UDPGT 3A1; UGT3A1 |
| Gene Symbols | UGT3A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.