Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT3A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563281 Supplier Invitrogen™ Supplier No. PA563281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSYQVIRWFS PEDHQKRIKK HFDSYIETAL DGRKESEALV KLMEIFGTQC SYLLSRKDIM Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 27%.

UDP-glucuronosyltransferases catalyze phase II biotransformation reactions in which lipophilic substrates are conjugated with glucuronic acid to increase water solubility and enhance excretion. They are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT3A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UGT3A1
Gene Accession No. Q6NUS8
Gene Alias FLJ26528; FLJ34658; UD3A1; UDP glycosyltransferase 3 family, polypeptide A1; UDP glycosyltransferase family 3 member A1; UDP-glucuronosyltransferase 3A1; UDPGT 3A1; UGT3A1
Gene Symbols UGT3A1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human UGT3A1 (Product #RP-100876)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.