Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UGT3A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565477
Description
Immunogen sequence: KSYQVIRWFSP EDHQKRIKKH FDSYIETALD GRKESEALVK LMEIFGTQCS YLLSRKDIM Highest antigen sequence identity to the following orthologs - mouse 45%, rat 27%.
UDP-glucuronosyltransferases catalyze phase II biotransformation reactions in which lipophilic substrates are conjugated with glucuronic acid to increase water solubility and enhance excretion. They are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds.
Specifications
UGT3A1 | |
Polyclonal | |
Unconjugated | |
UGT3A1 | |
FLJ26528; FLJ34658; UD3A1; UDP glycosyltransferase 3 family, polypeptide A1; UDP glycosyltransferase family 3 member A1; UDP-glucuronosyltransferase 3A1; UDPGT 3A1; UGT3A1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
133688 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NUS8 | |
UGT3A1 | |
Recombinant Human UGT3A1. Recombinant protein control fragment (Product #RP-100876). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction