Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UGT3A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563732
Description
Immunogen sequence: IPLFGDQPEN MVRVEAKKFG VSIQLKKLKA ETLALKMKQI M Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 63%.

Specifications
UGT3A2 | |
Polyclonal | |
Unconjugated | |
UGT3A2 | |
AI313915; PSEC0073; RGD1564365; UDP glucuronosyltransferase 3 family, polypeptide A2; UDP glycosyltransferase 3 family, polypeptide A2; UDP glycosyltransferase family 3 member A2; UDP glycosyltransferases 3 family, polypeptide A2; UDP-glucuronosyltransferase 3A2; UDPGT 3A2; Ugt3a2; UNQ842/PRO1780 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
167127 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3SY77 | |
UGT3A2 | |
Recombinant protein corresponding to Human UGT3A2. Recombinant protein control fragment (Product #RP-101857). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction