Learn More
Invitrogen™ UGT3A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596734
Description
Immunogen sequence: AKILTISTVG GSHYLLMDRV SQILQDHGHN VTMLNHKRGP FMPDFKKEEK SYQVISWLAP EDHQREFKKS FDFFLEETLG GRGKFENLLN VLEYLALQCS HFLNRKDIMD SLKNENFDMV IVETFDYCPF LIAEKLGKPF VAILSTSFGS LEFGLPIPLS YVPVFRSLLT DHMDFWGRVK NFLMFFSFCR RQQHMQSTFD NTIKEHFTEG SRPVLSHLLL KAELWFINSD FAFDFARPLL PNTVYVGGLM EKPIKPVPQD LENFIAKFGD SGFVLVTL; Positive Samples: B-cell, Jurkat, HepG2, Mouse kidney, Mouse spleen, Rat liver, Rat spleen; Cellular Location: Membrane, Single-pass type I membrane protein.

Specifications
UGT3A2 | |
Polyclonal | |
Unconjugated | |
UGT3A2 | |
AI313915; PSEC0073; RGD1564365; UDP glucuronosyltransferase 3 family, polypeptide A2; UDP glycosyltransferase 3 family, polypeptide A2; UDP glycosyltransferase family 3 member A2; UDP glycosyltransferases 3 family, polypeptide A2; UDP-glucuronosyltransferase 3A2; UDPGT 3A2; Ugt3a2; UNQ842/PRO1780 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
167127, 223337, 294793 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3SY77, Q8JZZ0 | |
UGT3A2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 23-300 of human UGT3A2 (NP_7775742). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.