Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553160
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553160 Supplier Invitrogen™ Supplier No. PA553160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SALHERGHHT VFLLSEGRDI APSNHYSLQR YPGIFNSTTS DAFLQSKMRN IFSGRLTAIE LFDILDHYTK NCDLMVGNHA LIQGLKKEKF DLLLVDPNDM CGFVIAHLLG VKYAVFSTGL WYPAEVGAPA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

UGT8 (UDP-galactose-ceramide galactosyltransferase, 2-hydroxyacylsphingosine 1-beta-galactosyltransferase) is a 541 amino acid, single pass membrane protein of the UDP-glycosyltransferase family. UGT8 is believed to be primarily involved with the metabolism of sphingolipids and galactosylceramide biosynthesis. UGT8 is one of six genes whose elevated expression has been correlated with a significantly increased the risk of lung metastases in breast cancer patients. As such, UGT8 may be a significant index of tumor aggressiveness and a potential marker for the prognostic evaluation of lung metastases in breast cancer. UGT8 is ubiquitously expressed with highest levels found in central and peripheral nervous systems and is up-regulated in breast cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ugt8
Gene Accession No. Q16880
Gene Alias 2-hydroxyacylsphingosine 1-beta-galactosyltransferase; AI850488; AW455908; ceramide UDP-galactosyltransferase; Cerebroside synthase; Cgt; mCerGT; UDP galactosyltransferase 8; UDP galactosyltransferase 8A; UDP glycosyltransferase 8; UDP-galactose ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase 8; UDP-galactose-ceramide galactosyltransferase 8A; UDP-glucuronosyltransferase 8; Ugt4; UGT8; Ugt8a; uridine diphosphate glycosyltransferase 8
Gene Symbols Ugt8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UGT8. Recombinant protein control fragment (Product #RP-90684).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7368
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.