Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UGT8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118340
This item is not returnable.
View return policy
Description
UGT8 Polyclonal Antibody for Western Blot, IP
UGT8 (UDP-galactose-ceramide galactosyltransferase, 2-hydroxyacylsphingosine 1-beta-galactosyltransferase) is a 541 amino acid, single pass membrane protein of the UDP-glycosyltransferase family. UGT8 is believed to be primarily involved with the metabolism of sphingolipids and galactosylceramide biosynthesis. UGT8 is one of six genes whose elevated expression has been correlated with a significantly increased the risk of lung metastases in breast cancer patients. As such, UGT8 may be a significant index of tumor aggressiveness and a potential marker for the prognostic evaluation of lung metastases in breast cancer. UGT8 is ubiquitously expressed with highest levels found in central and peripheral nervous systems and is up-regulated in breast cancers.
Specifications
| UGT8 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q16880 | |
| Ugt8 | |
| E. coli-derived Human UGT8 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Ugt8 | |
| 2-hydroxyacylsphingosine 1-beta-galactosyltransferase; AI850488; AW455908; ceramide UDP-galactosyltransferase; Cerebroside synthase; Cgt; mCerGT; UDP galactosyltransferase 8; UDP galactosyltransferase 8A; UDP glycosyltransferase 8; UDP-galactose ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase; UDP-galactose-ceramide galactosyltransferase 8; UDP-galactose-ceramide galactosyltransferase 8A; UDP-glucuronosyltransferase 8; Ugt4; UGT8; Ugt8a; uridine diphosphate glycosyltransferase 8 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7368 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction