Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UHMK1 Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene
SDP

Catalog No. 502542070
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2070 30 μL
50-167-2838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2070 Supplier OriGene Supplier No. TA501719S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UHMK1 Monoclonal antibody specifically detects UHMK1 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen UHMK1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2D3
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RBBP9
Gene Accession No. O75884
Gene Alias 4732477C12Rik; 4930500M09Rik; AA673513; AI449218; AU021979; kinase interacting stathmin; kinase interacting with leukemia-associated gene (stathmin); kinase interacting with stathmin; KIS; KIS protein kinase; KIST; PAM COOH-terminal interactor protein 2; P-CIP2; Serine/threonine-protein kinase Kist; U2AF homology motif (UHM) kinase 1; U2AF homology motif kinase 1; Uhmk1
Host Species Mouse
Immunogen Full length human recombinant protein of human RBBP9 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10741
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.