Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UHMK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113209
Description
UHMK1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Upon serum stimulation, KIS, a member of the Ser/Thr protein kinase family, phosphorylates CDKN1B/p27Kip1, thus controlling CDKN1B subcellular location and cell cycle progression in G1 phase. This protein, which contains 1 RRM (RNA recognition motif)domain, may be involved in trafficking and/or processing of RNA. KIS is widely expressed, with highest levels in skeletal muscle, kidney, placenta and peripheral blood leukocytes.
Specifications
| UHMK1 | |
| Polyclonal | |
| Unconjugated | |
| UHMK1 | |
| 4732477C12Rik; 4930500M09Rik; AA673513; AI449218; AU021979; kinase interacting stathmin; kinase interacting with leukemia-associated gene (stathmin); kinase interacting with stathmin; KIS; KIS protein kinase; KIST; PAM COOH-terminal interactor protein 2; P-CIP2; Serine/threonine-protein kinase Kist; U2AF homology motif (UHM) kinase 1; U2AF homology motif kinase 1; Uhmk1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 127933, 16589 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P97343, Q8TAS1 | |
| UHMK1 | |
| Recombinant Human Serine/threonine-protein kinase Kist protein (183-385AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction