Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UHRF1 Antibody (HL2287) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325732
Description
UHRF1 Monoclonal antibody specifically detects UHRF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
UHRF1 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HL2287 | |
Western Blot 1:1000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
E3 ubiquitin-protein ligase UHRF1, EC 6.3.2, EC 6.3.2.-, FLJ21925, hNP95, huNp95, ICBP90NP95, Inverted CCAAT box-binding protein of 90 kDa, Np95, Nuclear protein 95, Nuclear zinc finger protein Np95, RING finger protein 106, RNF106MGC138707, Transcription factor ICBP90, Ubiquitin-like PHD and RING finger domain-containing protein 1, ubiquitin-like with PHD and ring finger domains 1, ubiquitin-like, containing PHD and RING finger domains, 1, Ubiquitin-like-containing PHD and RING finger domains protein 1 | |
Recombinant fragment of human UHRF1. | |
100 μL | |
DNA Repair, Zinc Finger | |
29128 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction