Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UHRF1 Monoclonal Antibody (1A1B1)
GREENER_CHOICE

Catalog No. PIMA538654
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538654 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538654 Supplier Invitrogen™ Supplier No. MA538654
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2 and M phases of the cell cycle. It plays a major role in the G1/S transition by regulating topoisomerase IIalpha and retinoblastoma gene expression, and functions in the p53-dependent DNA damage checkpoint. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UHRF1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1A1B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene UHRF1
Gene Accession No. Q96T88
Gene Alias Ac2-121; AL022808; E3 ubiquitin-protein ligase UHRF1; fb97f09; FLJ21925; hNP95; Hnp95 Huhrf1; hUHRF1; HuNp95; ICBP90; inverted CCAAT box-binding protein of 90 kDa; Liver regeneration-related protein LRRG126; MGC138707; mUhrf1; Np95; nuclear phosphoprotein 95; nuclear protein; nuclear protein 95; Nuclear zinc finger protein Np95; RING finger protein 106; RING-type E3 ubiquitin transferase UHRF1; RNF106; TDRD22; transcription factor ICBP90; ub; ubiquitin like with PHD and ring finger domains 1; ubiquitin-like PHD and RING finger domain-containing protein 1; ubiquitin-like with PHD and ring finger domains 1; ubiquitin-like, containing PHD and RING finger domains, 1; ubiquitin-like-containing PHD and RING finger domains protein 1; Uhrf1; unm b1115; unm_b1115; wu:fb97f09; zgc:63539
Gene Symbols UHRF1
Host Species Mouse
Immunogen Purified recombinant fragment of human UHRF1 (aa: 616-755) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29128
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.