Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uhrf1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501994079
Encompass
Change view
Click to view available options
Quantity:
10 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4079 200 μL
50-199-4080 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4079 Supplier Active Motif Supplier No. 39625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2 and M phases of the cell cycle. It plays a major role in the G1/S transition by regulating topoisomerase IIalpha and retinoblastoma gene expression, and functions in the p53-dependent DNA damage checkpoint. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Uhrf1
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene UHRF1
Gene Accession No. Q96T88
Gene Alias Ac2-121; AL022808; E3 ubiquitin-protein ligase UHRF1; FLJ21925; hNP95; Hnp95 Huhrf1; hUHRF1; HuNp95; ICBP90; Inverted CCAAT box-binding protein of 90 kDa; MGC138707; mUhrf1; Np95; nuclear phosphoprotein 95; Nuclear protein 95; Nuclear zinc finger protein Np95; RING finger protein 106; RING-type E3 ubiquitin transferase UHRF1; RNF106; TDRD22; Transcription factor ICBP90; ub; ubiquitin like with PHD and ring finger domains 1; Ubiquitin-like PHD and RING finger domain-containing protein 1; ubiquitin-like with PHD and ring finger domains 1; ubiquitin-like, containing PHD and RING finger domains, 1; Ubiquitin-like-containing PHD and RING finger domains protein 1; UHRF1
Gene Symbols UHRF1
Host Species Rabbit
Immunogen This Uhrf1 antibody was raised against a peptide corresponding to the N-terminal half of human Uhrf1.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29128
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.