Learn More
UHRF1 Recombinant Rabbit Monoclonal Antibody (1H2L41), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen 702830

Description
This antibody is predicted to react with Monkey, Dog, Rat. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2 and M phases of the cell cycle. It plays a major role in the G1/S transition by regulating topoisomerase IIalpha and retinoblastoma gene expression, and functions in the p53-dependent DNA damage checkpoint. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
UHRF1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q96T88 | |
UHRF1 | |
Protein corresponding to human UHRF1 [126-285] | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1H2L41 | |
Unconjugated | |
UHRF1 | |
Ac2-121; AL022808; E3 ubiquitin-protein ligase UHRF1; fb97f09; FLJ21925; hNP95; Hnp95 Huhrf1; hUHRF1; HuNp95; ICBP90; inverted CCAAT box-binding protein of 90 kDa; Liver regeneration-related protein LRRG126; MGC138707; mUhrf1; Np95; nuclear phosphoprotein 95; nuclear protein; nuclear protein 95; Nuclear zinc finger protein Np95; RING finger protein 106; RING-type E3 ubiquitin transferase UHRF1; RNF106; TDRD22; transcription factor ICBP90; ub; ubiquitin like with PHD and ring finger domains 1; ubiquitin-like PHD and RING finger domain-containing protein 1; ubiquitin-like with PHD and ring finger domains 1; ubiquitin-like, containing PHD and RING finger domains, 1; ubiquitin-like-containing PHD and RING finger domains protein 1; Uhrf1; unm b1115; unm_b1115; wu:fb97f09; zgc:63539 | |
Rabbit | |
Protein A | |
RUO | |
29128 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.