Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UHRF1BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556929 Supplier Invitrogen™ Supplier No. PA556929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VQSEALAPDS MSHPRSKTEH DLKSLSGLTE VMEILKEGSS GMDNKGPLTE LEDVADVHML VHSPAHVRVR LDHYQYLALL RLKEVLQRLQ EQLTKDTESM TGSP Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.

UHRF1BP1 (UHRF1 binding protein 1) is a 1,440 amino acid protein that interacts with UHRF1 and may act as a negative regulator of cell growth. The UHRF1BP1 protein has been identified as a possible risk loci for systemic lupus erythematosus. The UHRF1BP1 gene is conserved in chimpanzee, dog, cow, mouse, rat, chicken, zebrafish and C.elegans, and maps to human chromosome 6p21.31.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UHRF1BP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UHRF1BP1
Gene Accession No. Q6BDS2
Gene Alias 1110020K19Rik; AA408674; C6orf107; dJ349A12.1; F830021D11Rik; ICBP90; ICBP90 binding protein 1; ICBP90-binding protein 1; mKIAA4127; Ubiquitin-like containing PHD and RING finger domains 1-binding protein 1; UHRF1 (ICBP90) binding protein 1; UHRF1 binding protein 1; UHRF1-binding protein 1; Uhrf1bp1
Gene Symbols UHRF1BP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UHRF1BP1. Recombinant protein control fragment (Product #RP-94622).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54887
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.