Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UHRF1BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572698
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572698 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572698 Supplier Invitrogen™ Supplier No. PA572698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UHRF1BP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

UHRF1BP1 (UHRF1 binding protein 1) is a 1,440 amino acid protein that interacts with UHRF1 and may act as a negative regulator of cell growth. The UHRF1BP1 protein has been identified as a possible risk loci for systemic lupus erythematosus. The UHRF1BP1 gene is conserved in chimpanzee, dog, cow, mouse, rat, chicken, zebrafish and C.elegans, and maps to human chromosome 6p21.31.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UHRF1BP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene UHRF1BP1
Gene Accession No. Q6BDS2
Gene Alias 1110020K19Rik; AA408674; C6orf107; dJ349A12.1; F830021D11Rik; ICBP90; ICBP90 binding protein 1; ICBP90-binding protein 1; mKIAA4127; Ubiquitin-like containing PHD and RING finger domains 1-binding protein 1; UHRF1 (ICBP90) binding protein 1; UHRF1 binding protein 1; UHRF1-binding protein 1; Uhrf1bp1
Gene Symbols UHRF1BP1
Host Species Rabbit
Immunogen UHRF1BP1 antibody for a 17 amino acid synthetic peptide near the center of human UHRF1BP1. The immunogen is within amino acids 890 - 940 of UHRF1BP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224648, 54887
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.