Learn More
UHRF1BP1L/KIAA0701 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304945A

Description
The recommended shelf life for this product is 1 year from date of receipt.
UHRF1BP1L gene ontology annotations related to this gene include GARP complex binding; protein homodimerization activity.Specifications
UHRF1BP1L/KIAA0701 | |
Polyclonal | |
Unconjugated | |
UHRF1BP1L | |
KIAA0701 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23074 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
A0JNW5 | |
UHRF1BP1L | |
Between 1414 and 1464 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.