Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UHRF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110547 Supplier Invitrogen™ Supplier No. PA5110547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSSLPSTSKQ NDAQVKPSSH NPPKVKKTAR GGSSSQPSTS ARTCLIDPGF GLYKVNELVD ARDVGLGAWF EAHIHSVTRA SDGHSRGKTP LKNGSSYKRT NGNVNHNSKE NTNKLDNVPS TSNSDSVAAD EDVIYHIEYD EYPESGILEM NVKDLRPRAR TILKWNELNV GDVVMVNYNV ENPGKRGFWY DAEITTLKTI SRTKKEVRVK VFLGGSEGTL NDCRVMSVDE IFKIEKPGAH PISFADGKFL RKNDPECDLC GGDPDKTCHM CSCHKCGEKR DPNM.

This gene encodes a nuclear protein which is involved in cell-cycle regulation. The encoded protein is a ubiquitin-ligase capable of ubiquinating PCNP (PEST-containing nuclear protein), and together they may play a role in tumorigenesis. The encoded protein contains an NIRF_N domain, a PHD finger, a set- and ring-associated (SRA) domain, and a RING finger domain and several of these domains have been shown to be essential for the regulation of cell proliferation. This protein may also have a role in intranuclear degradation of polyglutamine aggregates. Alternative splicing results in multiple transcript variants some of which are non-protein coding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UHRF2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UHRF2
Gene Accession No. Q7TMI3
Gene Alias 2310065A22Rik; AI426270; AW214556; D130071B19Rik; E3 ubiquitin-protein ligase UHRF2; Nirf; np95/ICBP90-like RING finger protein; Np95-like ring finger protein; Nuclear protein 97; nuclear zinc finger protein Np97; RING finger protein 107; RING-type E3 ubiquitin transferase UHRF2; RNF107; RP11-472F14.2; TDRD23; ubiquitin like with PHD and ring finger domains 2; ubiquitin-like PHD and RING finger domain-containing protein 2; ubiquitin-like with PHD and ring finger domains 2, E3 ubiquitin protein ligase; ubiquitin-like, containing PHD and RING finger domains 2; ubiquitin-like, containing PHD and RING finger domains, 2; ubiquitin-like-containing PHD and RING finger domains protein 2; UHRF2; URF2
Gene Symbols UHRF2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 76-359 of mouse Uhrf2 (NP_659122.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109113, 309331
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.