Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ UK 370106
GREENER_CHOICESDP

Catalog No. 290010 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-001-0 Supplier R&D Systems™ Supplier No. 2900/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly selective MMP-3 and MMP-12 inhibitor

Highly selective MMP-3 and MMP-12 inhibitor (IC50 values are 0.023, 0.042, 1.75, 2.3, 5.8, 30.4, 34.2 and 66.9 μM at MMP 3, 12, 8, 13, 7, 9, 2 and 14 respectively.) Inhibits fibronectin cleavage (IC50 = 320 nM) and has little effect on keratinocyte migration in vitro. Substantially inhibits MMP-3 in an ex vivo model of chronic dermal ulcers.

Chemical Identifiers

CAS 230961-21-4
Molecular Formula C35H44N2O5

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 572.73
Purity >95%
Molecular Weight (g/mol) 572.746
Chemical Name or Material UK 370106
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.