Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UK 5099
GREENER_CHOICE

Catalog No. 418610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-861-0 10 mg
41-865-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-861-0 Supplier Tocris Bioscience™ Supplier No. 4186/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

MCT inhibitor; also inhibits mitochondrial pyruvate carrier

Inhibitor of plasma membrane monocarboxylate transporters (MCTs) and the mitochondrial pyruvate carrier (MPC).

Chemical Identifiers

CAS 56396-35-1
Molecular Formula C18H11N2O2-

Specifications

Quantity 10 mg
Formula Weight 288.3
Purity >99%
Molecular Weight (g/mol) 287.298
Chemical Name or Material UK 5099
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.