Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UL13_HHV11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567985 Supplier Invitrogen™ Supplier No. PA567985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UL13_HHV11 Polyclonal Antibody for Western Blot

UL13 is a multifunctional serine/threonine kinase that plays a role in several processes including egress of virus particles from the nucleus, modulation of the actin cytoskeleton and regulation of viral and cellular gene expression. Regulates the nuclear localization of viral envelopment factors UL34 and UL31, by phosphorylating the US3 kinase, indicating a role in nuclear egress. Disrupts host nuclear lamins, including LMNA and LMNB1. Phosphorylates the viral Fc receptor composed of glycoproteins E (gE) and I (gI). Phosphorylation of glycoprotein E (gE) by UL13 alters its subcellular localization, from the host early endosome to the plasma membrane. Participates in the transcriptional regulation of cellular and viral mRNAs mainly by phosphorylating the viral transcriptional regulator ICP22. Additional substrates have been identified, including UL41, UL49 or host EF1D.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UL13_HHV11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Host Species Rabbit
Immunogen Synthesized peptide derived from human UL13_HHV11.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.