Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UL49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144441 Supplier Invitrogen™ Supplier No. PA5144441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UL49 Polyclonal Antibody for Western Blot, ELISA

UL49 is a gene that is found in the genome of certain viruses, specifically in the family Herpesviridae. UL49 is a nonessential gene, meaning that it is not essential for the replication and survival of the virus, but it may play a role in viral pathogenesis and host immune evasion. The specific function of UL49 can vary depending on the virus in which it is found. For example, in herpes simplex virus type 1 (HSV-1), UL49 encodes a glycoprotein that is involved in the viral entry process. In other herpesviruses, the exact function of UL49 may be different.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UL49
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.851 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias UL49; Uncharacterized protein UL49
Host Species Rabbit
Immunogen Recombinant Human cytomegalovirus (strain AD169) (HHV-5) (HCMV) Uncharacterized protein UL49 (354-570aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.