Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-1 Antibody (13C8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | ULBP-1 |
---|---|
Clone | 13C8 |
Dilution | Western Blot 1:500-1:2000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
ULBP-1 Monoclonal antibody specifically detects ULBP-1 in Human samples. It is validated for Western Blot, ELISASpecifications
ULBP-1 | |
Western Blot 1:500-1:2000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1 | |
A synthesized peptide derived from Human ULBP-1 [UniProt Q9BZM6] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
13C8 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Immunology | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
80329 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title