Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-1 Antibody (237104), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2588K
Description
ULBP-1 Monoclonal antibody specifically detects ULBP-1 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
ULBP-1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
237104 | |
Flow Cytometry, CyTOF-ready | |
ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1 | |
BaF3 mouse pro-B cell line transfected with mouse ULBP-1/MULT-1 | |
0.1 mL | |
Immunology | |
80329 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction