Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-2/5/6 Antibody (165903) [DyLight 650], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1298W
Description
ULBP-2/5/6 Monoclonal antibody specifically detects ULBP-2/5/6 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOFSpecifications
ULBP-2/5/6 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
80328 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
165903 | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
ALCAN-alpha, N2DL2, NKG2DL2, RAET1H, RAET1L, UL16 binding protein 2, ULBP2 | |
BaF3 mouse pro-B cell line transfected with human ULBP-2, Accession # Q9BZM5 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction