Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-2 Antibody (022), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290060APC
Description
ULBP-2 Monoclonal antibody specifically detects ULBP-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ULBP-2 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
022 | |
Immunohistochemistry-Paraffin | |
ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2 | |
Rabbit immunized with purified, recombinant Human ULBP-2 (Uniprot#: Q9BZM5, Met1-Ser217) | |
0.1 mL | |
Immunology, Innate Immunity | |
80328 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction