Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    ULBP-3 Antibody (166510), HRP, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1517H
Description
ULBP-3 Monoclonal antibody specifically detects ULBP-3 in Human samples. It is validated for Flow Cytometry, Blocking Assay, CyTOFSpecifications
| ULBP-3 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Flow Cytometry, Blocking Assay, CyTOF | |
| 166510 | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| NKG2D ligand 3, RAET1N, UL16 binding protein 3 | |
| BaF3 mouse pro-B cell line transfected with human ULBP-3 | |
| 0.1 mL | |
| Immunology | |
| 79465 | |
| Store at 4C in the dark. | |
| IgG2a | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            