Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-3 Antibody (166510), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1517PEATT594
Description
ULBP-3 Monoclonal antibody specifically detects ULBP-3 in Human samples. It is validated for Flow CytometrySpecifications
ULBP-3 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
166510 | |
Flow Cytometry | |
NKG2D ligand 3, RAET1N, UL16 binding protein 3 | |
BaF3 mouse pro-B cell line transfected with human ULBP-3 | |
0.1 mL | |
Immunology | |
79465 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction