Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$382.00 - $646.00
Specifications
Antigen | ULBP-3 |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ULBP-3 Polyclonal specifically detects ULBP-3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ULBP-3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q9BZM4 | |
79465 | |
This antibody was developed against a recombinant protein corresponding to amino acids: AHSLWYNFTIIHLPRHGQQWCEVQSQVDQKNFLSYDCGSDKVLSMGHLEEQLYATDAWGKQLEM | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Immunology | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
NKG2D ligand 3, RAET1N, UL16 binding protein 3 | |
ULBP3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title