Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-4/RAET1E Antibody (709116) [Alexa Fluor™ 532], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6285X
Description
ULBP-4/RAET1E Monoclonal antibody specifically detects ULBP-4/RAET1E in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
ULBP-4/RAET1E | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
709116 | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
bA350J20.7, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E2, retinoic acid early transcript 1ERL-4, ULBP4MGC125308 | |
Mouse myeloma cell line NS0-derived recombinant human ULBP-4/RAET1E, Gly30-Asp225, Accession # Q8TD07 | |
0.1 mL | |
Immunology | |
135250 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction