Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-4/RAET1E Antibody (RAET1E 50/3), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250267R
Description
ULBP-4/RAET1E Monoclonal specifically detects ULBP-4/RAET1E in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| ULBP-4/RAET1E | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAET1E | |
| Recombinant His-tagged protein to human extra-cellular domain of ULBP-4/RAET1E, grown in E.coli | |
| 0.1 mL | |
| Immunology | |
| 135250 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA | |
| RAET1E 50/3 | |
| Western Blot, Flow Cytometry, ELISA | |
| bA350J20.7, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E2, retinoic acid early transcript 1ERL-4, ULBP4MGC125308 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction