Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-4/RAET1E Antibody (RAET1E 50/3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250267H
Description
ULBP-4/RAET1E Monoclonal specifically detects ULBP-4/RAET1E in Human samples. It is validated for Western Blot, ELISA.Specifications
ULBP-4/RAET1E | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
RAET1E | |
Recombinant His-tagged protein to human extra-cellular domain of ULBP-4/RAET1E, grown in E.coli | |
0.1 mL | |
Immunology | |
135250 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, ELISA | |
RAET1E 50/3 | |
Western Blot, ELISA | |
bA350J20.7, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E2, retinoic acid early transcript 1ERL-4, ULBP4MGC125308 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction