Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULBP-6/RAET1L Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306628APC
Description
ULBP-6/RAET1L Polyclonal antibody specifically detects ULBP-6/RAET1L in Human samples. It is validated for ELISASpecifications
| ULBP-6/RAET1L | |
| Polyclonal | |
| ELISA | |
| retinoic acid early transcript 1L protein, retinoic acid early transcript 1L, ULBP6 | |
| Produced in rabbits immunized with purified, recombinant Human ULBP-6/RAET1L (Accession#: AAK91503.1; Met1-Ser217) | |
| 0.1 mL | |
| Immunology | |
| 154064 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction